Learn More
Mouse anti-Rat IgG1/2a, Clone: G28-5, Secondary Antibody, BD
Marca: BD Biosciences 553879
Detalles adicionales : Peso : 0.01000kg
Descripción
The G28-5 antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. A suspension of pooled rat IgG1 was used as the source of immunogen.
Host Species: Mouse
Clone: G28-5
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Pooled rat IgG
ELISA, Flow Cytometry
Especificaciones
Mouse anti-Rat IgG1/2a | |
G28-5 | |
Mouse | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Affinity Purified | |
RUO | |
Secondary | |
IgG2a |