missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™

Mouse Polyclonal Secondary Antibody

Marca:  Invitrogen 31824

168.67 EUR hasta el 2024-03-29
Use el código promocional "21615" para aplicar la oferta.



Detalles adicionales : Peso : 0.00100kg

Código de producto. 10260444

  • 253.00€ / 1 ml
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31824 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31824 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with goat, human, mouse, and sheep serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.0 mL of distilled water. Country of Origin: USA

Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.
Especificaciones

Especificaciones

Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
Biotin
Mouse
1 mL
Secondary
4° C
IgG
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
1.8 mg/mL
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Affinity chromatography
RUO
Rabbit
Lyophilized
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales