Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody, Biotin, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen MH1515
Detalles adicionales : Peso : 0.50000kg
Descripción
Human IgG1 Fc Monoclonal antibody specifically detects Human IgG1 Fc in Human samples. It is validated for Flow Cytometry.Especificaciones
Human IgG1 Fc | |
Monoclonal | |
0.3-1 mg/mL | |
PBS with BSA and 5mM sodium azide; pH 7.5 | |
IgG gamma 1 | |
500 μL | |
Secondary | |
-20°C | |
IgG1 κ |
Flow Cytometry | |
HP6070 | |
Biotin | |
Mouse | |
purified | |
RUO | |
Human | |
Liquid |