missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Fc Secondary Antibody, HRP, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen SA135470
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody does not cross react with albumin fractions of human serum, human IgA, human IgM or albumin fractions from other species
Human IgG Fc Monoclonal antibody specifically detects Human IgG Fc in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Western Blot.Especificaciones
Human IgG Fc | |
Monoclonal | |
1.2 mg/mL | |
PBS with 50% glycerol, 5mg/mL BSA and no preservative; pH 7.5 | |
Purified human IgG. | |
1 mL | |
Secondary | |
-20°C | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
EFE 565 | |
HRP | |
Mouse | |
purified | |
RUO | |
Human | |
Liquid |