Learn More
Mouse anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
Mouse Polyclonal Secondary Antibody
Marca: Invitrogen 31107
Detalles adicionales : Peso : 0.80000kg
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31107 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31107 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, human and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Especificaciones
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Mouse | |
1.5 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.8 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Liquid |