Learn More
Mouse anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC, Invitrogen™
Mouse Polyclonal Secondary Antibody
Marca: Invitrogen 31512
Detalles adicionales : Peso : 0.00010kg
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Antibody Specificity: This antibody reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. This product exhibits minimal inherent cross-reactivity to mouse serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction to mouse, human and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Fluorophore: Fluorescein-5-isothiocyanate (FITC-isomer 1) Amax = 492 nm; Emax = 520 nm Fluorophore/Protein: 8.9 μg/mg; 3.3 moles FITC per mole IgG Restoration and Storage: Store product at 4°C until opened. Restore with 0.75 mL distilled water (1.5 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Especificaciones
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
FITC | |
Mouse | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
1.5 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Goat | |
Lyophilized |