missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse 5'-Nucleotidase/CD73 ELISA Kit
Description
Mouse 5'-Nucleotidase/CD73 quantitates mouse 5'-Nucleotidase/CD73 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse 5'-Nucleotidase/CD73.
CD73 (Ecto-5-prime-nucleotidase, 5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. CD73 consists of a dimer of 2 identical 70 kDa subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. CD73 is used as a marker of lymphocyte differentiation. Consequently, a deficiency of CD73 occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from CD73 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. The CD73 gene has been localized to 6q14-q21 by immunofluorescence and a study of a panel of human x mouse hybrids that contained fragments of chromosome 6 as translocations. Defects in the CD73 gene can lead to the calcification of joints and arteries, and intestinal tuberculosis. Two transcript variants encoding different isoforms of CD73 have been found.
Specifications
Specifications
| Accession Number | 2210401F01Rik; 5 nucleotidase, ecto; 5 ' nucleotidase, ecto; 5 '-NT; 5 '-nucleotidase; 5 '-nucleotidase ecto; 5 '-nucleotidase, ecto (CD73); AI447961; CALJA; CD73; E5NT; Ecto-5 '-nucleotidase; eN; eNT; NT; Nt5; Nt5e; Nte; Purine 5-Prime-Nucleotidase |
| Assay Range | 7 to 1,500 pg/mL |
| Assay Sensitivity | 7 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 23959 |
| Gene Symbol | NT5E |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?