missing translation for 'onlineSavingsMsg'
Learn More

MOSC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 24782-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813275

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

As a component of the benzamidoxime prodrug-converting complex required to reduce N-hydroxylated prodrugs, such as benzamidoxime. Also able to reduce N(omega)-hydroxy-L-arginine (NOHA) and N(omega)-hydroxy-N(delta)-methyl-L-arginine (NHAM) into L-arginine and N(delta)-methyl-L-arginine, respectively.
Especificaciones

Especificaciones

MOSC2
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q922Q1, Q969Z3
MOSC2, RP11 270A6.1
Rabbit
IgG
150 μL
-20°C
Primary
54996, 67247
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MARC2
Liquid
MARC2
MOSC2 Fusion Protein Ag20694
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados