Learn More
Micronova™ Mop Adapter Controlled Environment
Product Code. 13187626 Shop All Micronova Products
Change view
Click to view available options
Material:
Polypropylene, Stainless Steel (304)
Unit Size:
Each
Product Code. Material Unit Size
13187626 Polypropylene, Stainless Steel (304) Each
1 options
This item is not returnable. View return policy
Product Code. 13187626 Supplier Micronova™ Supplier No. QDSLRTT14P
  • ONLINE EXCLUSIVE PRICE
    266.00€
    252.72€ / paquete de 1
    Ahorro 13.28€
    4% Descuento
 Restricted Item
Fecha estimada de envoi: 01-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The Micronova TankTool Adapter is a stainless steel frame adapter with swivel action that holds MZSM series of replaceable mop heads. Curved frame maintains contact while cleaning tanks. Available in exterior and interior (reverse) configurations
inside tank for use with Slimline SLSC and SLP slip covers, stainless steel/polyethylene, fit Slimline and Slim T bucket systems Slimline
Specifications
For Use With (Equipment) Stainless Steel Frame Adapter with Swivel action that holds MZSM series of replaceable Mop Heads
Description TankTool Adapter
For Use With (Application) Designed for Cleanroom Use
Certifications/Compliance ISO 9001:2008
Material Polypropylene, Stainless Steel (304)