Learn More
Monomethyl Phthalate O-Beta-D-Glucuronide, TRC
Product Code. 30009888
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009888 5 mg 5mg
30006139 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009888 Supplier Toronto Research Chemicals Supplier No. TRCM5665455MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53819-80-0
Molecular Formula C15H16O10
Molecular Weight (g/mol) 356.28
Synonym 1-(Methyl 1,2-Benzenedicarboxylate) β-D-Glucopyranuronic Acid, Monomethylphthalate Glucuronide;
SMILES COC(=O)C1=CC=CC=C1C(=O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C15H16O10/c1-23-13(21)6-4-2-3-5-7(6)14(22)25-15-10(18)8(16)9(17)11(24-15)12(19)20/h2-5,8-11,15-18H,1H3,(H,19,20)/t8-,9-,10+,11-,15-/m0/s1
Chemical Name or Material Monomethyl Phthalate O-beta-D-Glucuronide