Learn More
Invitrogen™ Lissamine™ Rhodamine B 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (rhodamine DHPE)
Product Code. 11564956
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
11564956 5 mg 5mg
1 options
This item is not returnable. View return policy
Product Code. 11564956 Supplier Invitrogen™ Supplier No. L1392
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
With excitation/emission maxima ∽560/580nm
The phospholipid, rhodamine DHPE is labeled on the head group with the red-fluorescent rhodamine B fluorophore.
Cell Analysis, Cell Structure, Membranes (General) & Lipids
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mg
Physical Form Solid
Product Line Invitrogen
For Research Use Only. Not for use in diagnostic procedures.