Learn More
Invitrogen™ Image-IT™ LIVE Green Reactive Oxygen Species Detection Kit, for microscopy
Product Code. 10738994
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
10738994 1 Kit 1 set
1 options
This item is not returnable. View return policy
Product Code. 10738994 Supplier Invitrogen™ Supplier No. I36007
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For microscopy
  • Image-iT LIVE Green Reactive Oxygen Species (ROS) Detection Kit provides key reagents necessary for detection of ROS in live cells
  • Assay is based on 5-(and-6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate (carboxy-H2DCFDA), reliable fluorogenic marker for ROS in live cells
  • In addition to carboxy-H2DCFDA, kit provides common inducer of ROS production tert-butyl hydroperoxide (TBHP), as positive control and blue-fluorescent, cell-permeant nucleic acid stain Hoechst 33342
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Product Line Image-iT
Product Type ROS Detection Kit
For Research Use Only. Not for use in diagnostic procedures.