Learn More
Invitrogen™ Bis-ANS (4,4'-Dianilino-1,1'-Binaphthyl-5,5'-Disulfonic Acid, Dipotassium Salt)
Product Code. 11544157
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
11544157 10 mg 10mg
1 options
This item is not returnable. View return policy
Product Code. 11544157 Supplier Invitrogen™ Supplier No. B153
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Binds with affinity that is often orders-of-magnitude higher than 1-8-ANS as probe for nonpolar cavities in proteins
Anilinonaphthalene sulfonates (ANS) are essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes. This property makes it a sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.
Cell Analysis, Cell Structure, Cellular Imaging, Cytoskeleton, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids
Product Line Molecular Probes
Label Type Other Label(s) or Dye(s)
Molecular Formula C32H22K2N2O6S2
Physical Form Solid
Product Type Protein Folding Indicator
For Research Use Only. Not for use in diagnostic procedures.