Learn More
Ansell Model 213 - AlphaTec™ 3000 Apron
Product Code. 18823036 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
Product Code. Size
18823036 Universal
1 options
This item is not returnable. View return policy
Product Code. 18823036 Supplier Ansell Supplier No. YE30T0021300
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Durable, comfortable multi-layer chemical barrier.
  • Tie fastening to waist
  • 100 cm long tie fastening
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Limited-use
Closure Type Ties
Sterility Non-sterile