missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10374-2-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa. The gene is part of a cluster of MMP genes which localize to chromosome 11q22. 3.Especificaciones
MMP7 | |
Polyclonal | |
Unconjugated | |
MMP7 | |
Matrilysin, Matrin, Matrix metalloproteinase 7, MMP 7, MMP7, MPSL1, PUMP 1, Pump 1 protease, PUMP1, Uterine metalloproteinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17393, 4316 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09237, Q10738 | |
Mmp7 | |
MMP7 Fusion Protein Ag0550 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |