Learn More
MMP-7 Antibody (MM0022-4C21), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB110-60988AF594
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215065
  • 575.58€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
MMP-7 Monoclonal specifically detects MMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
MMP-7
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
MMP7
Human recombinant MMP-7
0.1 mL
Primary
This antibody recognizes human MMP-7 in western blot without cross reactivity with other human MMPs. Recognizes both pro- (28 kDa) and active-form (18 kDa).
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0022-4C21
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase
Mouse
Protein G purified
Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, Wnt Signaling Pathway
4316.0
Human
IgG2b