missing translation for 'onlineSavingsMsg'
Learn More

Mitofusin 2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-66383AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216144

  • 518.35€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Mitofusin 2 Polyclonal specifically detects Mitofusin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Mitofusin 2
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CMT2A, CMT2A2, CPRP1mitochondrial assembly regulatory factor, EC 3.6.5, EC 3.6.5.-, HSG, KIAA0214hyperplasia suppressor, MARF, mitofusin 2, mitofusin-2, Transmembrane GTPase MFN2
Rabbit
Affinity Purified
Mitochondrial Markers
9927.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
MFN2
Partial recombinant human MFN2 protein (amino acids 364-599). [O95140]
0.1 mL
Primary
Human, Mouse, Rat
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales