missing translation for 'onlineSavingsMsg'
Learn More

Mitofusin 1 Antibody (11E9-1H12), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-71775R

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213931

  • 572.34€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Mitofusin 1 Monoclonal specifically detects Mitofusin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Especificaciones

Especificaciones

Mitofusin 1
Monoclonal
DyLight 550
DKFZp762F247, EC 3.6.5, EC 3.6.5.-, FLJ20693, Fzo homolog, hfzo1, hfzo2, MGC41806, mitochondrial transmembrane GTPase Fzo-1, mitochondrial transmembrane GTPase FZO-2, mitofusin 1, mitofusin-1, putative transmembrane GTPase, Transmembrane GTPase MFN1
Mouse
80 kDa
0.1 mL
Cancer, Cell Biology, Cellular Markers, Mitochondrial Fusion Proteins, Mitochondrial Markers, Neurodegeneration, Neuroscience, Signal Transduction
55669
Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
11E9-1H12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
MFN1
Mouse Mitofusin 1 [Swiss-Prot: Q811U4]
Protein G purified
RUO
Primary
Human and Mouse. Rat reactivity reported in scientific liteRature (PMID: 23658809).
Store at 4C in the dark.
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only