missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12214-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MINA is nuclear localized, myc-inducible protein that is thought to play a role in mammalian cell proliferation. Treatment of cancer cells lines such as the colon cancer cell line SW680 with siRNA against MINA inhibits cell growth, demonstrating that MINA may be a potential therapeutic target. MINA regulates several genes related to cell adhesion and metabolism that have also been shown to be regulated by c-Myc, but also regulates other genes whose expression are not modulated by c-Myc such as EGFR, IL-6 and HGF. MINA has also been found to act as a repressor to IL-4 expression in T cells, indicating that it may also play a role in T cell differentiation and genetic variation in T helper type 2 bias.Especificaciones
MINA | |
Polyclonal | |
Unconjugated | |
RIOX2 | |
MDIG, MINA, Mina53, MYC induced nuclear antigen, NO52, Nucleolar protein 52 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
266670, 67014, 84864 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CD15, Q8CFC1, Q8IUF8 | |
RIOX2 | |
MINA Fusion Protein Ag2838 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |