missing translation for 'onlineSavingsMsg'
Learn More

MIG6/ERRFI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11630-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818843

  • 109.00€ / 20 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995 ). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000 ; Fiorentino et al., 2000 ).
Especificaciones

Especificaciones

MIG6/ERRFI1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05432, Q99JZ7, Q9UJM3
ERRFI1, GENE 33, MIG 6, MIG6, RALT
Rabbit
IgG
20 μL
-20°C
Primary
313729, 54206, 74155
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
ERRFI1
Liquid
Errfi1
MIG6; ERRFI1 Fusion Protein Ag2202
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados