Learn More
Applied Biosystems™ MicroAmp™ GPLE Adhesive Film Applicators
Product Code. 15206555 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
5 Applicators
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
15206555 5 Applicators Pack of 5
1 options
This item is not returnable. View return policy
Product Code. 15206555 Supplier Applied Biosystems™ Supplier No. 4481196
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Applied Biosystems™ MicroAmp™ GPLE Adhesive Film Applicators form a tight seal between a microplate and an adhesive film. Each lot of these General Purpose Laboratory Equipment (GPLE) applicators is manufactured in an ISO-registered facility.
Use an Applied Biosystems MicroAmp GPLE Adhesive Film Applicator to form a tight seal between a microplate and an adhesive film. Each lot of these General Purpose Laboratory Equipment (GPLE) applicators is manufactured in an ISO-registered facility.
Form a tight seal
The MicroAmp GPLE Adhesive Film Applicator enables you to ergonomically apply pressure to an adhesive film upon a microplate to form a tight seal and minimize air bubbles. This tight seal can help prevent well-to-well contamination and sample loss.
Quick and easy
Use the MicroAmp GPLE Adhesive Film Applicator to speed the process of sealing multiple microplates. This tool is designed according to the microplate dimensions so you can quickly seal a microplate. The rugged proprietary plastic endures constant use so you can complete large screening applications.
Note: See user's manual or package insert for limited label license and trademark information.
TRUSTED_SUSTAINABILITY
Specifications
Product Line MicroAmp
Color Gray
Material Polyethylene
No. per Pack 5 Applicators
Product Type Film, Foil, or Seal Applicator
Quantity 5 Applicators
For Laboratory Use