Learn More
Emma Safety Footwear Micro Safety Shoes
Unique microfiber upper is fully washable. SmartecAir™ provides fresh dry feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors used in high-tech and cleanroom environments.
Marca:  Emma Safety Footwear 500540-47
Detalles adicionales : Peso : 1.54300kg
 Ver más versiones de este producto
Código de producto. 15928461
  • 75.05€ / 1 par
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Especificaciones
Especificaciones
EN ISO 20345:2011, EN 12568:2010
White
Hydro-Tec™ Sanitized Silver
47
Elastic
Micro
Microfiber
Steel Toe