Learn More
SI Analytics™ Micro Ostwald Viscometer
Product Code. 10778741 Shop All SI Analytics Products
Change view
Click to view available options
Approximate Constant:
0.01K
0.1K
Size:
0.43mm Diameter
0.77mm Diameter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Approximate Constant Size Unit Size
10778741 0.01K 0.43mm Diameter Each
10233551 0.1K 0.77mm Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778741 Supplier SI Analytics™ Supplier No. 285404203
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Determine absolute and relative kinematic viscosity of liquids with Newtonian flow behavior. SI Analytics™ Micro Ostwald Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, high or low temperature measurements, and minimum measurement substance or rinsing agent quantities
  • Calibrated
  • With ring marks
  • With constants
  • For manual measurements
  • Filling quantity of 2mL
  • Overall length of 290mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Calibrated Yes
Approximate Constant 0.01K
Includes Ring Mark
For Use With (Application) Measurements of Small Liquid Quantities Even With Extreme Formation of Foam
Size 0.43mm Diameter
Volume (Metric) 2 mL