missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MICA Human ELISA Kit

Sandwich ELISA Kit

Marca:  Invitrogen™ BMS2302TEN

Detalles adicionales : Peso : 1.90000kg

 Ver más versiones de este producto

Código de producto. 15924214

  • 3165.00€ / paquete de 10


Fecha estimada de envío
25-08-2020
Añadir a la cesta

Descripción

Descripción

Major histocompatibility complex (MHC) class I proteins are ubiquitously expressed and mediate the recognition of intracellular antigens by cytotoxic T cells. A related family of proteins, the MHC class I chain-related (MIC) proteins, are recognized by NKG2D, a receptor on NK and T cells, and promote anti-tumor activity. MICA, a member of the MIC family, is widely expressed on many tumors, and it is the MICA/NKG2D interaction that is thought to stimulate the anti-tumor reactivity by T lymphocytes. Both MICA and MICB mRNA are widely expressed in normal tissues, with MICA being present in virtually every tissue except the nervous system, suggesting that MIC protein expression may only be one component of the anti-tumor activity of the immune system.
Especificaciones

Especificaciones

Q29983
Biotin
Pre-coated 96 well plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers|
RUO
Serum, 50 μL|Plasma, 50 μL|Cell Lysate,50 μL|Supernatant, 50 μL
1 hr. 20 min.
Human
6.6%
96 x 10 Tests
Supernatant
Colorimetric Microplate Reader
1.6-100 pg/ml
100507436
ELISA Kit
1.09 pg/ml
2°C to 8°C
4 hr.
MICA
5.1%
Human
HRP
Videos
Documentos