Learn More
Metrizoic Acid, TRC
Product Code. 30477458
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30477458 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30477458 Supplier Toronto Research Chemicals Supplier No. TRCM338935500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Metrizoic Acid
CAS 1949-45-7
Molecular Formula C12H11I3N2O4
Quantity 500 mg
Synonym 3-(Acetylamino)-5-(acetylmethylamino)-2,4,6-triiodobenzoic Acid, 3-Acetamido-2,4,6-triiodo-5-(N-methylacetamido)benzoic Acid, 3,5-Diacetamido-N-methyl-2,4,6-triiodobenzoic Acid, Metrizoate
InChI Formula InChI=1S/C12H11I3N2O4/c1-4(18)16-10-7(13)6(12(20)21)8(14)11(9(10)15)17(3)5(2)19/h1-3H3,(H,16,18)(H,20,21)
SMILES CN(C(=O)C)c1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I
IUPAC Name 3-acetamido-5-[acetyl(methyl)amino]-2,4,6-triiodobenzoic acid
Molecular Weight (g/mol) 627.94
Formula Weight 627.7853