missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methylene dithiocyanate, 98%

Marca:  Alfa Aesar™ A10740.18

Detalles adicionales : CAS : 6317-18-6 Peso : 0.05000kg Transport : UN number : 2928 Chem class : 6.1 Pack group : II

Cantidad. 50g

 Ver más versiones de este producto

Código de producto. 11499612

  • 90.60€ / 50 g


Stock EU 1
Añadir a la cesta

Descripción

Descripción

Applications
Methylene dithiocyanate is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field. It is also used to control slime in pulp and paper manufacturing, preservation of paints, adhesives, synthetic polymer lattices and latex emulsions, anti-microbial treatment of water thinned products, thickeners and slurries, preservation of wood and wood products, preservation of Leather and Leather products and in oil well brines and drilling mud.

Solubility
Decomposes in water. Slightly soluble in organic solvents.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Especificaciones

Identificadores de productos químicos

6317-18-6
130.183
JWZXKXIUSSIAMR-UHFFFAOYSA-N
22772
C(SC#N)SC#N
C3H2N2S2
MFCD00001833
methylene dithiocyanate, methylene bis thiocyanate, methylenedithiocyanate, proxel mb, methylenedirhodanide, thiocyanic acid, methylene ester, nalfloc n 206, methylenedirhodanid, methylene thiocyanate, dithiocyanatomethane
thiocyanatomethyl thiocyanate

Especificaciones

Methylene dithiocyanate
103°C to 107°C
6317-18-6
MFCD00001833
methylene dithiocyanate, methylene bis thiocyanate, methylenedithiocyanate, proxel mb, methylenedirhodanide, thiocyanic acid, methylene ester, nalfloc n 206, methylenedirhodanid, methylene thiocyanate, dithiocyanatomethane
C(SC#N)SC#N
130.183
130.19
>110°C (230°F)
50g
C3H2N2S2
UN2928
JWZXKXIUSSIAMR-UHFFFAOYSA-N
thiocyanatomethyl thiocyanate
22772
98%
Videos
SDS