Learn More
Metil tricosanoato, 99+ %, Thermo Scientific Chemicals
Catalog Number p-4089644
Click to view available options
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
2433-97-8
C24H48O2
368.65
MFCD00009349
VORKGRIRMPBCCZ-UHFFFAOYSA-N
tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
75519
metil tricosanoato
CCCCCCCCCCCCCCCCCCCCCCC(=O)OC
2433-97-8
C24H48O2
368.65
MFCD00009349
VORKGRIRMPBCCZ-UHFFFAOYSA-N
tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
75519
metil tricosanoato
CCCCCCCCCCCCCCCCCCCCCCC(=O)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl tricosanoate
Punto de fusión 53°C
Temperatura de inflamación >113°C
Intervalo de porcentaje de ensayo 99% min. (GC)
Envase Frasco de vidrio
Fórmula lineal CH3(CH2)21CO2CH3
Cantidad 1 g
Formula Weight (peso de la fórmula) 368.64
Porcentaje de pureza 99+%
compliance-icons
Nombre del producto
  • Methyl tricosanoate
Palabra de advertencia
  • Peligro
Clasificación
  • Lesión ocular grave/irritación ocular Categoría 1