Learn More
Metilfenilpropiolato, 97 %, Thermo Scientific Chemicals
Catalog Number p-7027659
Click to view available options
Cantidad:
5 g
25 g
100 g
4891-38-7
C10H8O2
160.172
MFCD00041685
JFGWPXKGINUNDH-UHFFFAOYSA-N
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
138378
metil 3-fenilprop-2-inoato
COC(=O)C#CC1=CC=CC=C1
4891-38-7
C10H8O2
160.172
MFCD00041685
JFGWPXKGINUNDH-UHFFFAOYSA-N
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
138378
metil 3-fenilprop-2-inoato
COC(=O)C#CC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl phenylpropiolate
Densidad 1.083
Punto de ebullición 109°C to 110°C (2 mmHg)
Temperatura de inflamación >110°C (230°F)
Fórmula lineal C6H5C≡CCO2CH3
Índice de refracción 1.562
Cantidad 5 g
Beilstein 472813
Formula Weight (peso de la fórmula) 160.17
Porcentaje de pureza 97%