Learn More
Methyl o-tolylacetate, 98%, Thermo Scientific™
Product Code. 11312327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312327 5 g 5g
11322327 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09877.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl o-tolylacetate is used as pharmaceutical intermediates.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40851-62-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00082780
InChI Key BLEMRRXGTKTJGT-UHFFFAOYSA-N
Synonym methyl o-tolylacetate, methyl 2-2-methylphenyl acetate, methyl 2-o-tolylacetate, methyl 2-o-tolyl acetate, methyl 2-methylphenylacetate, methyl 2-methylphenyl-acetate, methyl 2-methylphenyl acetate, methyl 2-methyl phenyl acetate, 2-methylbenzeneacetic acid methyl ester, 2-methylphenyl acetic acid methyl ester
PubChem CID 7021479
IUPAC Name methyl 2-(2-methylphenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1C
Specifications
Boiling Point 70°C to 71°C (1 mmHg)
Refractive Index 1.511
Quantity 5 g
Beilstein 2208739
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material Methyl o-tolylacetate