missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl diphenylphosphite

Marca:  Alfa Aesar™ 032097.03

Detalles adicionales : CAS : 3577-87-5 Peso : 0.00100kg Transport : UN number : 3278 Chem class : 6.1 Pack group : III

Cantidad. 1g

 Ver más versiones de este producto

Código de producto. 11318327

  • 77.80€ / 1 g


En stock 3
Añadir a la cesta

Especificación

Identificadores de productos químicos

3577-87-5
248.218
NYCZNDFWFCCTPA-UHFFFAOYSA-N
520764
COP(OC1=CC=CC=C1)OC2=CC=CC=C2
C13H13O3P
MFCD00014886
diphenyl methyl phosphite, methyldiphenylphosphite, methoxydiphenoxyphosphine, methoxy-diphenoxy-phosphane, methyl diphenylphosphite 1g, phosphorous acid, methyl diphenyl ester
methyl diphenyl phosphite

Especificación

Methyl diphenylphosphite
1g
C13H13O3P
UN3278
NYCZNDFWFCCTPA-UHFFFAOYSA-N
methyl diphenyl phosphite
520764
Liquid
Hygroscopic
3577-87-5
MFCD00014886
diphenyl methyl phosphite, methyldiphenylphosphite, methoxydiphenoxyphosphine, methoxy-diphenoxy-phosphane, methyl diphenylphosphite 1g, phosphorous acid, methyl diphenyl ester
COP(OC1=CC=CC=C1)OC2=CC=CC=C2
248.218
248.21
Videos
SDS