Learn More
Metil dimetoxiacetato, + 97 %, Thermo Scientific Chemicals
Catalog Number p-7081961
Click to view available options
Cantidad:
5 g
10 g
50 g
250 g
89-91-8
C5H10O4
134.131
MFCD00008484
NZTCVGHPDWAALP-UHFFFAOYSA-N
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
66647
metil 2,2-dimetoxiacetato
COC(C(=O)OC)OC
89-91-8
C5H10O4
134.131
MFCD00008484
NZTCVGHPDWAALP-UHFFFAOYSA-N
methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
66647
metil 2,2-dimetoxiacetato
COC(C(=O)OC)OC
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl dimethoxyacetate
Densidad 1.098
Punto de ebullición 160°C to 163°C
Temperatura de inflamación 63°C (145°F)
Índice de refracción 1.405
Cantidad 5 g
Beilstein 1757582
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 134.13
Porcentaje de pureza ≥97%