Learn More
Methyl acetate, CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Catalog Number p-7122128
Click to view available options
Cantidad:
250 mL
2.5 L
79-20-9
C3H6O2
74.079
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
79-20-9
C3H6O2
74.079
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton, devoton, acetic acid, methyl ester, methyl ethanoate, acetic acid methyl ester, methylacetat, acetate de methyle, methylacetaat, methyl acetic ester, octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
CHROMASOLV™, for HPLC,≥99.8%
  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl acetate
Densidad 0.934g/mL at 25°C
Temperatura de inflamación −13°C (8.6°F)
Envase Glass bottle
Fórmula lineal CH3COOCH3
Cantidad 250 mL
Número UN UN1231
Beilstein 1736662
Densidad del vapor 2.55 (vs air)
Formula Weight (peso de la fórmula) 74.08g/mol
Porcentaje de pureza ≥99.8% (GC)∼∼≥99.8%