Learn More
Alfa Aesar™ Methyl 4-(triphenylphosphonio)crotonate bromide, 98%
Marca: Alfa Aesar™ A10443.06
Detalles adicionales : CAS : 2181-98-8 Peso : 0.00500kg
Cantidad. | 5g |
---|
Identificadores de productos químicos
2181-98-8 | |
441.305 | |
KSTXYAKHACCZSD-NWBUNABESA-M | |
6450201 | |
COC(=O)C=CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-] |
C23H22BrO2P | |
MFCD00031575 | |
methyl 4-triphenylphosphonio crotonate bromide, 4-methoxy-4-oxobut-2-en-1-yl triphenylphosphonium bromide, 2e-4-methoxy-4-oxobut-2-en-1-yl triphenylphosphanium bromide, 3-methoxycarbonyl-2-propenyl triphenylphosphonium bromide, 4-methoxy-4-oxo-2-butenyl triphenylphosphonium bromide, methyl 4-triphenylphosphonio crotonatebromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium bromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphonium bromide, z-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium,bromide, e-4-methoxy-4-oxidanylidene-but-2-enyl-triphenyl-phosphanium bromide | |
[(E)-4-methoxy-4-oxobut-2-enyl]-triphenylphosphanium;bromide |
Especificación
Methyl 4- (triphenylphosphonio)crotonate bromide | |
5g | |
C23H22BrO2P | |
4173385 | |
KSTXYAKHACCZSD-NWBUNABESA-M | |
[(E)-4-methoxy-4-oxobut-2-enyl]-triphenylphosphanium;bromide | |
6450201 | |
98% |
~175°C (decomposition) | |
2181-98-8 | |
MFCD00031575 | |
methyl 4-triphenylphosphonio crotonate bromide, 4-methoxy-4-oxobut-2-en-1-yl triphenylphosphonium bromide, 2e-4-methoxy-4-oxobut-2-en-1-yl triphenylphosphanium bromide, 3-methoxycarbonyl-2-propenyl triphenylphosphonium bromide, 4-methoxy-4-oxo-2-butenyl triphenylphosphonium bromide, methyl 4-triphenylphosphonio crotonatebromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium bromide, e-4-methoxy-4-oxobut-2-enyl-triphenylphosphonium bromide, z-4-methoxy-4-oxobut-2-enyl-triphenylphosphanium,bromide, e-4-methoxy-4-oxidanylidene-but-2-enyl-triphenyl-phosphanium bromide | |
COC(=O)C=CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-] | |
441.305 | |
441.32 |