missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1-(p-toluenosulfonil)pirrol-2-carboxilato de metilo, 95 %, Thermo Scientific™

Catalog Number p-7032119
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
930111-86-7
C13H12N2O6S
324.31
MFCD09743415
AYFBDJWRDWDJJZ-UHFFFAOYSA-N
methyl 1-4-methylbenzenesulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-tosyl-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-p-toluenesulfonyl pyrrole-2-carboxylate, methyl 1-4-methylphenyl sulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-toluene-4-sulphonyl-1h-pyrrole-2-carboxylate, methyl 1-4-methylbenzene-1-sulfonyl-4-nitro-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-4-methylphenyl sulphonyl-1h-pyrrole-2-carboxylate
44784923
1-(4-metilfenil)sulfonil-4-nitropirrol-2-carboxilato de metilo
COC(=O)C1=CC(=CN1S(=O)(=O)C1=CC=C(C)C=C1)[N+]([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
930111-86-7
C13H12N2O6S
324.31
MFCD09743415
AYFBDJWRDWDJJZ-UHFFFAOYSA-N
methyl 1-4-methylbenzenesulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-tosyl-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-p-toluenesulfonyl pyrrole-2-carboxylate, methyl 1-4-methylphenyl sulfonyl-4-nitropyrrole-2-carboxylate, methyl 4-nitro-1-toluene-4-sulphonyl-1h-pyrrole-2-carboxylate, methyl 1-4-methylbenzene-1-sulfonyl-4-nitro-1h-pyrrole-2-carboxylate, methyl 4-nitro-1-4-methylphenyl sulphonyl-1h-pyrrole-2-carboxylate
44784923
1-(4-metilfenil)sulfonil-4-nitropirrol-2-carboxilato de metilo
COC(=O)C1=CC(=CN1S(=O)(=O)C1=CC=C(C)C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl 4-nitro-1-(p-toluenesulfonyl)pyrrole-2-carboxylate
Punto de fusión 198°C to 200°C
Cantidad 250 mg
Formula Weight (peso de la fórmula) 260.25
Porcentaje de pureza 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'