missing translation for 'onlineSavingsMsg'
Learn More

4-Metoxipiridina-2-carboxilato de metilo, 98 %, Thermo Scientific Chemicals

Catalog Number 15441769
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
29681-43-4
C8H9NO3
167.164
MFCD00956223
OJDKENGKKYVJLY-UHFFFAOYSA-N
methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
1519345
4-metoxipiridina-2-carboxilato de metilo
COC1=CC(=NC=C1)C(=O)OC
Este artículo no se puede devolver. Vea la política de devoluciones
29681-43-4
C8H9NO3
167.164
MFCD00956223
OJDKENGKKYVJLY-UHFFFAOYSA-N
methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
1519345
4-metoxipiridina-2-carboxilato de metilo
COC1=CC(=NC=C1)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl 4-methoxypyridine-2-carboxylate
Punto de fusión 47°C to 50°C
Color Blanco a amarillo
Cantidad 1 g
Beilstein 473379
Formula Weight (peso de la fórmula) 167.16
Porcentaje de pureza 98%
Forma física Sólido

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'