Learn More
Methyl 3,5-Dichlorobenzoate, TRC
Product Code. 30134458
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134458 500 mg 500mg
30134570 1 g 1g
30003372 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134458 Supplier Toronto Research Chemicals Supplier No. TRCM326343500MG
 Restricted Item
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2905-67-1
Molecular Formula C8 H6 Cl2 O2
Molecular Weight (g/mol) 205.04
Synonym Benzoic acid, 3,5-dichloro-, methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Methyl 3,5-dichlorobenzoate
IUPAC Name methyl 3,5-dichlorobenzoate
SMILES COC(=O)c1cc(Cl)cc(Cl)c1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H6Cl2O2/c1-12-8(11)5-2-6(9)4-7(10)3-5/h2-4H,1H3
Formula Weight 203.9745
Chemical Name or Material 3,5-Dichlorobenzoic acid methyl ester