missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxocyclopentanecarboxylate, 97%

Product Code. 11400808
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400808 25 g 25g
11490798 5 g 5g
11410808 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400808 Supplier Thermo Scientific Alfa Aesar Supplier No. B25256.14
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy

CAS: 10472-24-9 | C7H10O3 | 142.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10472-24-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00001411
InChI Key PZBBESSUKAHBHD-UHFFFAOYSA-N
Synonym methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane
PubChem CID 66328
IUPAC Name methyl 2-oxocyclopentane-1-carboxylate
SMILES COC(=O)C1CCCC1=O

Specifications

Density 1.145
Boiling Point 105°C to 107°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4565
Quantity 25 g
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material Methyl 2-oxocyclopentanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.