Learn More
Metil 2-metilbutirato, 98 %, Thermo Scientific Chemicals
Catalog Number 15294804
Click to view available options
Cantidad:
100 g
1 kg
5 kg
868-57-5
C6H12O2
116.16
MFCD00009335
OCWLYWIFNDCWRZ-UHFFFAOYNA-N
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
13357
metil 2-metilbutanoato
CCC(C)C(=O)OC
868-57-5
C6H12O2
116.16
MFCD00009335
OCWLYWIFNDCWRZ-UHFFFAOYNA-N
methyl 2-methylbutyrate, butanoic acid, 2-methyl-, methyl ester, methyl 2-methyl butyrate, methyl s-2-methylbutanoate, 2-methylbutyric acid methyl ester, butyric acid, 2-methyl-, methyl ester, dl-2-methylbutyric acid methyl ester, methyl dl-2-methylbutyrate, methyl alpha-methylbutyrate, methyl alpha-methylbutanoate
13357
metil 2-metilbutanoato
CCC(C)C(=O)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Methyl 2-methylbutyrate
Densidad 0.8850g/mL
Punto de ebullición 115.0°C
Temperatura de inflamación 18°C
Intervalo de porcentaje de ensayo 97.5% min. (GC)
Envase Tambor de plástico
Fórmula lineal CH3CH2CH(CH3)COOCH3
Cantidad 5 kg
Gravedad específica 0.885
Información de solubilidad Solubility in water: practically insoluble
Formula Weight (peso de la fórmula) 116.16
Porcentaje de pureza 98%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Methyl 2-methylbutyrate
Palabra de advertencia
  • Peligro
Clasificación
  • Lesión ocular grave/irritación ocular Categoría 2