Learn More
Metapneumovirus Nucleoprotein Antibody (hMPV123), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NBP2-53104AF350
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215774
  • 509.71€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
Metapneumovirus Nucleoprotein Monoclonal specifically detects Metapneumovirus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.
Especificaciones
Especificaciones
Metapneumovirus Nucleoprotein
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Human MPV strain NCL-145
0.1 mL
Primary
Virus
IgG2a
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
hMPV123
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
Virology Bacteria and Parasites
Identification and location of human metapneumovirus (hMPV) of both types A and B.
Store at 4°C in the dark.