missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Photometer Tube Test

Product Code. 11933787
Change view
Click to view available options
Range:
0 to 2 mg/L
0 to 20 mg/L
0 to 7 mg/L Zn, 0 to 35 mg/L Zn
0.2 to 20 mg/L
0.3 to 20 mg/L
No. of Tests:
25
30
Unit Size:
Pack of 25
Pack of 30
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range No. of Tests unitSize
11933787 0.2 to 20 mg/L 25 Pack of 25
11988987 0 to 2 mg/L 30 Pack of 30
11998987 0 to 20 mg/L 30 Pack of 30
11943787 0.3 to 20 mg/L 25 Pack of 25
11963787 0 to 7 mg/L Zn, 0 to 35 mg/L Zn 25 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11933787 Supplier Palintest™ Supplier No. PL427

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Predispensed reagent tests require minimal handling and speed up processing. Palintest™; Photometer Tube Test reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.

  • For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
  • For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
  • Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.

Specifications

Analytes Copper
For Use With (Equipment) Photometer, heater block
No. of Tests 25
Quantity 1 Each
Range 0.2 to 20 mg/L
Type Photometer tube test reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.