Learn More
Pacific Handy Cutter Metal Detectable Safety Cutter
Product Code. 17798967
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
Product Code. Color Unit Size
17798967 Blue Pack of 50
1 options
This item is not returnable. View return policy
Product Code. 17798967 Supplier Pacific Handy Cutter Supplier No. DFC364MD
  • ONLINE EXCLUSIVE PRICE
    513.33€
    422.41€ / paquete de 50
    Ahorro 90.92€
    17% Descuento
Fecha estimada de envoi: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The DFC364MD is a metal detectable safety cutter with a concealed blade. It offers the highest level of security and thus protects people and goods at the same time.
  • Ideal for use in areas where metal detectability is important, e.g. the food, chemical or pharmaceutical industries.
  • Our film specialist - particularly suitable for puncturing due to the long nose.
  • Designed for foil of any kind, adhesive tape, banding & strapping.
  • The integrated tape splitter protects the blade.
  • Disposable product.
  • With practical eyelet for lanyard fastening or hanging.
Specifications
Product Type Metal Detectable Knife
Disposable Single-use
Sterility Non-sterile
Color Blue