missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meta-Phosphoric Acid, ACS Reagent, Honeywell Fluka™
Click to view available options
Cantidad:
5 g
100 g
500 g
2.5 kg
6 x 500 g
Especificaciones
Nombre del producto químico o material | Meta-Phosphoric Acid |
Concentración | 33.5 to 36.5% |
Cantidad | 2.5 kg |
Fórmula lineal | (HPO3)n |
Número UN | 3260 |
Grado | ACS Reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'