Learn More
Trajan Scientific and Medical™ MEPS BINs for Agilent 7693A Syringes
Miniaturization of conventional sample preparation devices. Trajan Scientific and Medical MEPS BINs for Agilent 7693A Syringes is micro extraction by packed sorbent.
Marca:  Trajan Scientific and Medical™ 2900601
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15289233
  • 479.00€ / paquete de 5
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Save hours in sample preparation
  • Reduce solvent consumption and sample volume
  • Available in reversed phase, normal phase, mixed mode and ion exchange chemistries
Especificaciones
Especificaciones
Gas chromatography
Cone Tipped Needle
C18
GPC Applications
Videos