missing translation for 'onlineSavingsMsg'
Learn More

Mena Antibody (ENAH/1988), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79887AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213516

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

Mena
Monoclonal
Alexa Fluor 594
50mM Sodium Borate with 0.05% Sodium Azide
ENAH
Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) (Uniprot: Q8N8S7)
0.1 mL
Cancer, Neuroscience
Human
IgG2c κ
Western Blot, ELISA
ENAH/1988
Western Blot, ELISA
ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales