Learn More
MP Biomedicals™ Membranes for Filtration Unit for SafTest™ Test Kits
Product Code. 15681874 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
18 Pk.
Unit Size:
Pack of 18
Product Code. Quantity Unit Size
15681874 18 Pk. Pack of 18
1 options
This item is not returnable. View return policy
Product Code. 15681874 Supplier MP Biomedicals™ Supplier No. 07DP1000
  • ONLINE EXCLUSIVE PRICE
    819.00€
    780.15€ / paquete de 18
    Ahorro 38.85€
    4% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Membranes for Filtration Unit for SafTest™ solid sample testing platform.
  • Replacement filters for use with the SafTest sample testing platform.
  • Includes 18 manifolds of 6 filters each.
Specifications
For Use With (Application) Food Quality Testing
Quantity 18 Pk.
Storage Requirements Room temperature