Learn More
DWK Life Sciences DURAN™ Membrane Screw Cap
Venting membrane improves autoclaving safety and storage of sterile liquids in GL32 bottles and erlenmeyers
Marca:  DWK Life Sciences 291185609
Detalles adicionales : Peso : 0.01900kg
 Ver más versiones de este producto
Código de producto. 15876921
  • 27.60€ / paquete de 5
Fecha de envío estimada
del proveedor 07-11-2018
Añadir a la cesta
Descripción
Descripción
Reduce contamination risk in autoclaving, as the 0.2 um membrane permits pressure equalization with screw tight sealing. Ingress of liquids or solids is prevented and the bottle contents remain sterile.
Especificaciones
Especificaciones
White
Autoclaving sterile liquids, pressure equilization applications
Yes
40mm
0.2μm membrane size
Polypropylene, PTFE membrane
GL 56
Videos