missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT14 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26953-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
MCT14 | |
Polyclonal | |
Unconjugated | |
SLC16A14 | |
MCT 14, MCT14, Monocarboxylate transporter 14, SLC16A14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100519114, 151473, 71781 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7RTX9, Q8K1C7 | |
Slc16a14 | |
MCT14 Fusion Protein Ag25315 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG |