missing translation for 'onlineSavingsMsg'
Learn More

MCM6 Antibody (MCM6/2999), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79862V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214238

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

MCM6 Monoclonal specifically detects MCM6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

MCM6
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
MCM6
Recombinant fragment (around aa 228-368) of human MCM6 protein (exact sequence is proprietary) (Uniprot: Q14566)
0.1 mL
Cell Cycle and Replication, DNA Repair
Human
IgG2b κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
MCM6/2999
Immunohistochemistry, Immunohistochemistry-Paraffin
DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales