Learn More
Cole-Parmer™ MasterflexLive™ L/S Cloud-Enabled Digital Drive
Monitor and control your pump from anywhere and at any time when using your MasterflexLive-equipped pump drive! Using cloud-based technology, you can monitor and control your pump using your smart phone as an interface, freeing you from being physically tethered to your application.
Marca:  Cole-Parmer™ 07551-60
Detalles adicionales : Peso : 7.71000kg
 Ver más versiones de este producto
Código de producto. 15923572
  • 3415.18€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • MasterflexLive is a secure, cloud-enabled platform for control and monitoring of Masterflex pumps
  • Get real-time control of all pump parameters – including speed, flow rate, dispense volume, and more
  • Push notifications alert you to operating conditions and error messages
  • Monitor and adjust critical processes running 24/7 without the need for onsite personnel
  • Internet connection via standard Ethernet (RJ45) jack on rear of drive
  • Flow rate 0.001 to 588 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually
  • Open-head sensor stops the drive when the pump head is opened (when used with sensor-equipped pump head)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
Especificaciones
Especificaciones
IP33 per IEC 60529; Conforms to ANSI/UL Std 61010-1 Certified to CAN/CSA Std C22.2 No. 61010-1. This product has been tested to the requirements of CAN/CSA-C22.2 No. 61010-1, second edition, including Amend- ment 1, or a later version of the same standard incorporating the same level of testing requirements. (For CE Mark): EN61010-1 (EU Low Voltage Directive) and EN61326 (EU EMC Directive)
±0.1% (0.01 rpm @ 100 rpm)
White w/accents
267 x 203 x 203mm
ABS Plastic, aluminum, vinyl
90 to 260 Vrms @ 50/60 Hz
6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country specific plug/cord set; please specify ultimate destination when ordering.
40°C
32°F
Accepts up to 4
0.02-100 rpm
Masterflex L/S Tubing
Masterflex Live 07551-60 L/S Cloud-Enabled Digital Drive, 0.02 to 100 rpm, 115/230 VAC
No
(Universal Input) Single Phase Only (Universal Input) Single Phase Only
ABS Plastic and Aluminum
128 x 64 LCD w/ LED Backlight
Easy-Load II w/ Open-Head Sensor, Easy-Load II, Easy-Load, Standard, High-Performance, PTFE Tubing, PTFE Diaphragm pump heads
IP33
Brushless DC
0°C
Yes
Digital Variable Speed
5.9 kg
Videos