missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14829-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MARS is a member of aminoacyl-tRNA synthetases which are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene belongs to the class I family of tRNA synthetases.Especificaciones
MARS | |
Polyclonal | |
Unconjugated | |
MARS | |
MARS, Methionine tRNA ligase, methionyl tRNA synthetase, METRS, MTRNS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216443, 299851, 4141 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56192, Q68FL6 | |
MARS | |
MARS Fusion Protein Ag6619 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |